Learn More
WTW™ Measuring Vessel
WTW offers a wide variety of measuring bottles and measuring vessels with adapters to measure BSB, soil respiration, degradeability as per OECD 301F, fermentation tests, biogas and much more. They have all be tested in their respective applications and therefore support accurate measurements.
Brand: WTW™ 209023
2576.12 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Measuring Vessel | |
Rack and Seal, 2 x GK 600 Rubber Sleeve, 4 x DV/MG/KL Clamps and Beaker MB 50 (50mL) |
Product Content Correction
Product Title
WTW™ Measuring Vessel > Includes: Rack and Seal, 2 x GK 600 Rubber Sleeve, 4 x DV/MG/KL Clamps and Beaker MB 50 (50mL)
Spot an opportunity for improvement?Share a Content Correction