missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meclizine dihydrochloride, MP Biomedicals™
Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Color | White |
Quantity | 25 g |
Formula Weight | 463.9 |
Physical Form | Powder |
Chemical Name or Material | Meclizine dihydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'