missing translation for 'onlineSavingsMsg'
Learn More

AMPri™ Med-Comfort Premium Grip Latex Examination Gloves, PPE Cat. III

Catalog Number 16846081
Click to view available options
Size:
Large
Medium
Small
X-Large
X-Small
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-sterile AMPri™ Med-Comfort latex examination gloves are powder-free and have a textured finish for good grip. Ideal for general laboratory use as well as clinical, pharmaceutical and food use applications, they offer excellent protection against microorganisms (viral, bacteria and fungi).

  • AQL 1.5
  • Premium grip examination gloves
  • Applications include laboratory, clinical, pharmaceutical, dental, and food industry use
  • Protect against chemicals and microorganisms
  • Ambidextrous fit
  • Powder-free
  • Non-sterile
  • Color: white
  • Textured finish
  • Disposable
Certifications/Compliance AQL 1.5
Certifications/Compliance (Europe) EN 455-1, EN374-1/4/5, EN420, Food grade
Color White
Disposable Single-use
Laboratory/Cleanroom Use Laboratory
Latex Free No
Size X-Small
Thickness (Metric) 0.08 mm (Cuff)
Length (Metric) 240 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'