Learn More
Systec™ MediaPrep-10
For preparing and sterilizing microbiological media. Systec MediaPrep-10 is equipped and controlled to be used as normal autoclave.
Brand:  Systec™ 1710
Product Code. 12022949
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Locking of door and loading hatch is dependent on temperature and pressure
  • Accidental opening is not possible
  • Inner vessel holder is exchangeable and easy to remove and clean
  • Powerful heating elements enable rapid heating
  • Rapid cooling is by water circulation and support presssure directly at external walls of media vessel using sterilized demineralized water
  • Water is brought to cooling temperature using plate heat exchanger with water connection
  • Total process time including heating, sterilizing and cooling to 50°C lasts from 60–120 minutes, depending on size of vessel, quantity of media and temperature of cooling water
  • Support pressure avoids overboiling respectively foaming of media
  • Air compressor is built-in
Specifications
Specifications
16 A
11.6 in.
Preparing and sterilizing microbiological media
220 - 240 V (single phase), 50/60 Hz, 16 A
222 mm Inner Vessel
8.7 in. Inner Vessel
3.6 kw
17 L
296 mm
MediaPrep
250 mm
9.8 in.
50/60 Hz
550 x 655 x 530 mm (21.6 x 25.7 x 20.8 in.)