Learn More
Gibco™ MEM, Hanks' Balanced Salts
MEM, Hanks' Balanced Salts
255.00 SEK - 2075.00 SEK
Specifications
Certifications/Compliance | ISO 13485 standards, FDA |
---|---|
Content And Storage | 2 to 8°C |
Form | Liquid |
Product Type | MEM Hanks' Balanced Salts |
With Additives | Hanks’ Balanced Salt solution (HBSS), Glutamine, Phenol Red |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11540576
|
Gibco™
21575097 |
10 x 500 mL |
2075.00 SEK
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11530576
|
Gibco™
21575022 |
500 mL |
255.00 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
Dual-Site cGMP Manufacturing
Gibco™ MEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). This MEM formulation does not require a CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
Liquid | |
Hanks’ Balanced Salt solution (HBSS), Glutamine, Phenol Red | |
Mammalian cell culture |
2 to 8°C | |
MEM Hanks' Balanced Salts | |
No HEPES | |
For in-vitro diagnostic use |
Safety and Handling
missing translation for 'shelfLife' : 12 Months