missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
216.00 SEK - 369.00 SEK
Chemical Identifiers
CAS | 7783-34-8 |
---|---|
Molecular Formula | H2HgN2O7 |
Molecular Weight (g/mol) | 342.62 |
MDL Number | MFCD00149736 |
InChI Key | KVICROHOONHSRH-UHFFFAOYSA-N |
PubChem CID | 3084029 |
IUPAC Name | mercury(2+) hydrate dinitrate |
SMILES | O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Description
Chemical Identifiers
7783-34-8 | |
342.62 | |
KVICROHOONHSRH-UHFFFAOYSA-N | |
mercury(2+) hydrate dinitrate |
H2HgN2O7 | |
MFCD00149736 | |
3084029 | |
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
7783-34-8 | |
H2HgN2O7 | |
Liquid | |
1 | |
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
342.62 |
HDPE plastic bottle | |
MFCD00149736 | |
Colorless | |
KVICROHOONHSRH-UHFFFAOYSA-N | |
mercury(2+) hydrate dinitrate | |
3084029 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Mercury solution 1000 ppm
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mercury Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™