missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress [Met5]-Enkephalin, amide TFA
MedChemExpress [Met5]-Enkephalin, amide TFA is an agonist for δ opioid receptors as well as putative ζ (zeta) opioid receptors.
Brand: MedChemExpress HY-P1467A
This item is not returnable.
View return policy
Specifications
98.35% | |
Neuroscience-Neuromodulation | |
Peptides | |
Room temperature | |
5-Methionine-enkephalin amide TFA |
Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) | |
686.7 g/mol | |
25 mg | |
H2O : 100 mg/mL (145.62 mM∣Need ultrasonic) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction