missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aroclor 1248 in DCM 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
Specifications
Concentration | 100μg/mL |
CAS | 12672-29-6 |
Packaging | Ampoule |
Quantity | 1 mL |
Type | Organic Single-component Standard Solution |
Molecular Formula | C12H6Cl4 |
InChI Key | UTMWFJSRHLYRPY-UHFFFAOYSA-N |
SMILES | C1=C(C=C(C=C1Cl)Cl)C2=CC(=CC(=C2)Cl)Cl |
IUPAC Name | 1,3-dichloro-5-(3,5-dichlorophenyl)benzene |
Molecular Weight (g/mol) | 291.98 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'