missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metabolite Decabromodiphenyl ether in n-Nonane 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule(s)
Specifications
Specifications
Concentration | 100mu;g/mL |
CAS | 1163-19-5 |
Packaging | Ampoule(s) |
Quantity | 1 mL |
Molecular Formula | C12Br10O |
InChI Key | WHHGLZMJPXIBIX-UHFFFAOYSA-N |
SMILES | C1(=C(C(=C(C(=C1Br)Br)Br)Br)Br)OC2=C(C(=C(C(=C2Br)Br)Br)Br)Br |
IUPAC Name | 1,2,3,4,5-pentabromo-6-(2,3,4,5,6-pentabromophenoxy)benzene |
Molecular Weight (g/mol) | 959.171 |
PubChem CID | 14410 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'