missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endrin ketone in Methanol 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
Specifications
Specifications
Concentration | 100μg/mL |
CAS | 53494-70-5 |
Packaging | Ampoule |
Quantity | 1 mL |
Type | Organic Single-component Standard Solution |
Molecular Formula | C12H8Cl6O |
InChI Key | IZHZFAQWVKBTSL-DNZAWBEXSA-N |
SMILES | C1C2C3C4C1C(=O)C2C5(C3(C(C4(C5Cl)Cl)(Cl)Cl)Cl)Cl |
Molecular Weight (g/mol) | 380.895 |
PubChem CID | 131880538 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction