missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Metribuzin in Acetone 10μg/mL, Fisher Chemical™

Product Code. 12940004
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule(s)
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Concentration 10μg/mL
CAS 21087-64-9
Packaging Ampoule(s)
Quantity 10 mL
Molecular Formula C8H14N4OS
MDL Number MFCD00055525
UN Number 1090
InChI Key FOXFZRUHNHCZPX-UHFFFAOYSA-N
SMILES CSC1=NN=C(C(=O)N1N)C(C)(C)C
IUPAC Name 4-amino-6-tert-butyl-3-(methylsulfanyl)-4,5-dihydro-1,2,4-triazin-5-one
Molecular Weight (g/mol) 214.29
PubChem CID 30479
ChEBI CHEBI:34846
Solvent or Matrix Acetone
Chemical Name or Material Metribuzin
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Metribuzin 10ug/ml in acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.