Learn More
Viking Optical™ Metal Case Hygrometer
Brand: Viking Optical™ 7203
This item is not returnable.
View return policy
Description
Metal case. Please note that all hair hygrometers require periodic recalibration, at approximately six month intervals.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.