Learn More
S Murray™ Metal Luer-Lock Syringe
Use these syringes for a variety of research applications. S Murray™ Metal Luer-Lock Syringe are graduated and feature metal luer fittings.
Brand:  S Murray™ G813/05
 View more versions of this product
Product Code. 12359229
  • 315.00 SEK / Each
Explore more special offers
You may only order up to the available stock. For more details call customer services on +46 31 352 32 00.
Description
Description
Use these syringes for a variety of research applications. S Murray™ Metal Luer-Lock Syringe are graduated and feature metal luer fittings.
  • Syringe with metal luer fitting
  • Fully interchangeable
Specifications
Specifications
CE
Metal Luer-lock Syringe
20 mL
Yes
Glass
1/Ea.
Luer Lock