Learn More
S Murray™ Metal Luer-Tip Syringe
Use these syringes for a variety of research applications. S Murray™ Metal Luer-Tip Syringe are graduated and feature metal luer fittings.
Brand: S Murray™ G812/06
This item is not returnable.
View return policy
Description
Use these syringes for a variety of research applications. S Murray™ Metal Luer-Tip Syringe are graduated and feature metal luer fittings.
- Syringe with metal luer fitting
- Fully interchangeable
Specifications
CE | |
Metal Luer-tip Syringe | |
50 mL | |
Yes |
Glass | |
1/Ea. | |
Luer Tip |