Learn More
Methanesulfonamide, 97+%, Thermo Scientific Chemicals
407.00 SEK - 6450.00 SEK
Chemical Identifiers
CAS | 3144-09-0 |
---|---|
Molecular Formula | CH5NO2S |
Molecular Weight (g/mol) | 95.12 |
MDL Number | MFCD00007940 |
InChI Key | HNQIVZYLYMDVSB-UHFFFAOYSA-N |
Synonym | methylsulfonamide, methane sulfonamide, methanesulphonamide, methansulfonamid, methyl sulfonamide, aminosulfane dioxide, methyl sulfonylamine, ch3so2nh2, mesylamine, methansulfonamide |
PubChem CID | 72879 |
IUPAC Name | methanesulfonamide |
SMILES | CS(N)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11478820
|
Thermo Scientific Alfa Aesar
A10885.09 |
10 g |
407.00 SEK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488820
|
Thermo Scientific Alfa Aesar
A10885.18 |
50 g |
1175.00 SEK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498820
|
Thermo Scientific Alfa Aesar
A10885.30 |
250 g |
3750.00 SEK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424243
|
Thermo Scientific Alfa Aesar
A10885.36 |
500 g |
6450.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methanesulfonamide will react with thionyl chloride, cyanates, carbon disulfide and ketones and aldehydes to prepare pharmaceuticals, brightening agent and other target molecules. It is also used in biological studies to predict binding affinity and binding mode of protein ligand complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethanesulfonamide will react with thionyl chloride, cyanates, carbon disulfide and ketones and aldehydes to prepare pharmaceuticals, brightening agent and other target molecules. It is also used in biological studies to predict binding affinity and binding mode of protein ligand complexes.
Solubility
Slightly soluble in DMSO and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
3144-09-0 | |
95.12 | |
HNQIVZYLYMDVSB-UHFFFAOYSA-N | |
72879 | |
CS(N)(=O)=O |
CH5NO2S | |
MFCD00007940 | |
methylsulfonamide, methane sulfonamide, methanesulphonamide, methansulfonamid, methyl sulfonamide, aminosulfane dioxide, methyl sulfonylamine, ch3so2nh2, mesylamine, methansulfonamide | |
methanesulfonamide |
Specifications
3144-09-0 | |
CH5NO2S | |
1740835 | |
Slightly soluble in DMSO and methanol. | |
CS(N)(=O)=O | |
95.12 | |
95.13 | |
Methanesulfonamide |
87°C to 93°C | |
MFCD00007940 | |
methylsulfonamide, methane sulfonamide, methanesulphonamide, methansulfonamid, methyl sulfonamide, aminosulfane dioxide, methyl sulfonylamine, ch3so2nh2, mesylamine, methansulfonamide | |
HNQIVZYLYMDVSB-UHFFFAOYSA-N | |
methanesulfonamide | |
72879 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 221-553-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Methanesulfonamide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Methanesulfonamide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.