missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-methylcyclopropyl ketone, 95%, Thermo Scientific Chemicals

Product Code. 11904841
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1567-75-5
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
InChI Key OQBCJXUAQQMTRW-UHFFFAOYSA-N
Synonym methyl 1-methylcyclopropyl ketone, 1-1-methylcyclopropyl ethan-1-one, 1-1-methylcyclopropyl ethanone, 1-acetyl-1-methylcyclopropane, ethanone, 1-1-methylcyclopropyl, 1-methylcyclopropyl methyl ketone, methyl 1-methyl cyclopropyl ketone, 1-1-methyl-cyclopropyl-ethanone, methyl1-methylcyclopropylketone
PubChem CID 74067
IUPAC Name 1-(1-methylcyclopropyl)ethanone
SMILES CC(=O)C1(CC1)C

Specifications

Color Colorless to Yellow
Density 0.8950g/mL
Boiling Point 125.0°C to 128.0°C
Flash Point 24°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula CH3C3H4COCH3
Refractive Index 1.4330 to 1.4350
Quantity 100 g
Specific Gravity 0.895
Solubility Information Solubility in water: insoluble.
Formula Weight 98.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 1-methylcyclopropyl ketone
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 1-methylcyclopropyl ketone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'