missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-(2-hydroxyphenyl)propionate, 97%, Thermo Scientific Chemicals
687.54 SEK - 2934.77 SEK
Chemical Identifiers
CAS | 20349-89-7 |
---|---|
Molecular Formula | C10H12O3 |
Molecular Weight (g/mol) | 180.203 |
MDL Number | MFCD00067757 |
InChI Key | YHXYRISRGHSPNV-UHFFFAOYSA-N |
Synonym | methyl 3-2-hydroxyphenyl propanoate, methyl 3-2-hydroxyphenyl propionate, methyl melilotate, acmc-20aki4, methyl hydroxy-benzenepropionate, methyl 2'-hydroxyhydrocinnamate, methyl 3-hydroxyphenyl propionate, methyl 3 2-hydroxyphenyl propanoate, 2-hydroxybenzenepropanoic acid methyl ester, 3-2-hydroxyphenyl propionic acid methyl ester |
PubChem CID | 2794569 |
IUPAC Name | methyl 3-(2-hydroxyphenyl)propanoate |
SMILES | COC(=O)CCC1=CC=CC=C1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11464893
|
Thermo Scientific Alfa Aesar
A12219.06 |
5 g |
687.54 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474893
|
Thermo Scientific Alfa Aesar
A12219.14 |
25 g |
2934.77 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20349-89-7 | |
180.203 | |
YHXYRISRGHSPNV-UHFFFAOYSA-N | |
2794569 | |
COC(=O)CCC1=CC=CC=C1O |
C10H12O3 | |
MFCD00067757 | |
methyl 3-2-hydroxyphenyl propanoate, methyl 3-2-hydroxyphenyl propionate, methyl melilotate, acmc-20aki4, methyl hydroxy-benzenepropionate, methyl 2'-hydroxyhydrocinnamate, methyl 3-hydroxyphenyl propionate, methyl 3 2-hydroxyphenyl propanoate, 2-hydroxybenzenepropanoic acid methyl ester, 3-2-hydroxyphenyl propionic acid methyl ester | |
methyl 3-(2-hydroxyphenyl)propanoate |
Specifications
20349-89-7 | |
C10H12O3 | |
methyl 3-2-hydroxyphenyl propanoate, methyl 3-2-hydroxyphenyl propionate, methyl melilotate, acmc-20aki4, methyl hydroxy-benzenepropionate, methyl 2'-hydroxyhydrocinnamate, methyl 3-hydroxyphenyl propionate, methyl 3 2-hydroxyphenyl propanoate, 2-hydroxybenzenepropanoic acid methyl ester, 3-2-hydroxyphenyl propionic acid methyl ester | |
COC(=O)CCC1=CC=CC=C1O | |
180.203 | |
180.21 | |
Methyl 3- (2-hydroxyphenyl)propionate |
40°C to 44°C | |
MFCD00067757 | |
YHXYRISRGHSPNV-UHFFFAOYSA-N | |
methyl 3-(2-hydroxyphenyl)propanoate | |
2794569 | |
97% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only