missing translation for 'onlineSavingsMsg'
Learn More
Methyl 3-cyclopentenecarboxylate, 97%, Thermo Scientific Chemicals Product Code.: 15565966

Methyl 3-cyclopentenecarboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15565966
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15565966

Brand: Thermo Scientific Acros 457400010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58101-60-3
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD04038661
InChI Key CEOILRYKIJRPBZ-UHFFFAOYSA-N
Synonym methyl 3-cyclopentenecarboxylate, methyl cyclopent-3-enecarboxylate, methyl-3-cyclopentene-1-carboxylate, methyl3-cyclopentenecarboxylate, 3-cyclopentene-1-carboxylic acid methyl ester, 3-cyclopentene-1-carboxylic acid, methyl ester, acmc-209m2j, methyl cyclopent-3-ene carboxylate, 4-methoxycarbonyl cyclopent-1-ene, methyl 3-cyclopentene-1-carboxylate
PubChem CID 1514118
IUPAC Name methyl cyclopent-3-ene-1-carboxylate
SMILES COC(=O)C1CC=CC1

Specifications

Boiling Point 149.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 126.16
Percent Purity 97%
Chemical Name or Material Methyl 3-cyclopentenecarboxylate
compliance-icons compliance-icons
Product Identifier
  • Methyl 3-cyclopentenecarboxylate, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 3-cyclopentenecarboxylate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.