missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methoxy-4-nitrobenzoate, 98%, Thermo Scientific Chemicals

769.00 SEK - 2835.00 SEK

Chemical Identifiers

CAS 5081-37-8
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.173
MDL Number MFCD00051967
InChI Key PVVFEPFLFHDHHK-UHFFFAOYSA-N
Synonym 3-methoxy-4-nitrobenzoic acid methyl ester, benzoic acid, 3-methoxy-4-nitro-, methyl ester, benzoic acid, 5-methoxy-4-nitro-, methyl ester, methyl3-methoxy-4-nitrobenzoate, pubchem4216, acmc-209kq5, ksc275c3p, methylmethoxynitrobenzenecarboxylate, 4-nitro-3-methoxy benzoic acid, 3-methoxy-4-nitro-benzoic acid methyl ester
PubChem CID 287308
IUPAC Name methyl 3-methoxy-4-nitrobenzoate
SMILES COC1=C(C=CC(=C1)C(=O)OC)[N+](=O)[O-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11420797
SDS View Documents
Thermo Scientific Alfa Aesar
B23196.03
1 g N/A N/A N/A
11430797
SDS View Documents
Thermo Scientific Alfa Aesar
B23196.06
5 g N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 3-methoxy-4-nitrobenzoate, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.