Learn More
Methyl 3-(methylthio)propionate, 98%, Thermo Scientific Chemicals
380.00 SEK - 3385.00 SEK
Chemical Identifiers
CAS | 13532-18-8 |
---|---|
Molecular Formula | C5H10O2S |
Molecular Weight (g/mol) | 134.19 |
MDL Number | MFCD00008565 |
InChI Key | DMMJVMYCBULSIS-UHFFFAOYSA-N |
Synonym | methyl 3-methylthio propionate, methyl 3-methylthiopropionate, methyl 3-methylthio propanoate, propanoic acid, 3-methylthio-, methyl ester, methyl beta-methiopropionate, methyl 3-methylsulfanyl propanoate, methyl-3-methylthio propionate, methyl 3-methylthiopropanoate, propionic acid, 3-methylthio-, methyl ester, methyl beta-methylthiopropionate |
PubChem CID | 61641 |
IUPAC Name | methyl 3-methylsulfanylpropanoate |
SMILES | COC(=O)CCSC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11403394
|
Thermo Scientific Alfa Aesar
A10649.14 |
25 g |
380.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11413394
|
Thermo Scientific Alfa Aesar
A10649.22 |
100 g |
885.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423394
|
Thermo Scientific Alfa Aesar
A10649.36 |
500 g |
3385.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13532-18-8 | |
134.19 | |
DMMJVMYCBULSIS-UHFFFAOYSA-N | |
61641 | |
COC(=O)CCSC |
C5H10O2S | |
MFCD00008565 | |
methyl 3-methylthio propionate, methyl 3-methylthiopropionate, methyl 3-methylthio propanoate, propanoic acid, 3-methylthio-, methyl ester, methyl beta-methiopropionate, methyl 3-methylsulfanyl propanoate, methyl-3-methylthio propionate, methyl 3-methylthiopropanoate, propionic acid, 3-methylthio-, methyl ester, methyl beta-methylthiopropionate | |
methyl 3-methylsulfanylpropanoate |
Specifications
13532-18-8 | |
72°C to 73°C (11 mmHg) | |
C5H10O2S | |
1.465 | |
1745077 | |
DMMJVMYCBULSIS-UHFFFAOYSA-N | |
methyl 3-methylsulfanylpropanoate | |
61641 | |
98% |
1.074 | |
72°C (161°F) | |
CH3SCH2CH2CO2CH3 | |
MFCD00008565 | |
methyl 3-methylthio propionate, methyl 3-methylthiopropionate, methyl 3-methylthio propanoate, propanoic acid, 3-methylthio-, methyl ester, methyl beta-methiopropionate, methyl 3-methylsulfanyl propanoate, methyl-3-methylthio propionate, methyl 3-methylthiopropanoate, propionic acid, 3-methylthio-, methyl ester, methyl beta-methylthiopropionate | |
COC(=O)CCSC | |
134.19 | |
134.2 | |
Methyl 3- (methylthio)propionate |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 236-883-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.