Learn More
Methyl 3-oxocyclobutanecarboxylate, 97%, Thermo Scientific Chemicals
Description
Methyl 3-oxocyclobutanecarboxylate can be used as an intermediate for the synthesis of various pharmaceutical compounds. It is used for the preparation of novel imidazobenzazepine derivatives as dual H1/5-HT2A antagonists for the treatment of sleep disorders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl 3-oxocyclobutanecarboxylate can be used as an intermediate for the synthesis of various pharmaceutical compounds. It is used for the preparation of novel imidazobenzazepine derivatives as dual H1/5-HT2A antagonists for the treatment of sleep disorders.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Specifications
Specifications
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 128.12 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 3-oxocyclobutanecarboxylate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.