Learn More
Methyl 4-bromopyridine-2-carboxylate, 97+%, Thermo Scientific Chemicals
593.00 SEK - 9390.00 SEK
Chemical Identifiers
CAS 29681-42-3
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD07374897
InChI Key JZFLATQBIPILFS-UHFFFAOYSA-N
PubChem CID 12828947
IUPAC Name methyl 4-bromopyridine-2-carboxylate
SMILES COC(=O)C1=NC=CC(=C1)Br
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15471479
SDS View Documents
Thermo Scientific Alfa Aesar
H64327.03
1 g
593.00 SEK
1g
15481479
SDS View Documents
Thermo Scientific Alfa Aesar
H64327.06
5 g
2255.00 SEK
5g
15491479
SDS View Documents
Thermo Scientific Alfa Aesar
H64327.14
25 g
9390.00 SEK
25g
Description
Description
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.
Solubility
Slightly soluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications
Chemical Identifiers
29681-42-3
216.034
JZFLATQBIPILFS-UHFFFAOYSA-N
methyl 4-bromopyridine-2-carboxylate
C7H6BrNO2
MFCD07374897
12828947
COC(=O)C1=NC=CC(=C1)Br
Specifications
29681-42-3
C7H6BrNO2
473047
Slightly soluble in water.
COC(=O)C1=NC=CC(=C1)Br
216.034
216.03
Solid
54°C to 56°C
MFCD07374897
methyl 4-bromopicolinate, 4-bromo-pyridine-2-carboxylic acid methyl ester, methyl 4-bromo-2-pyridinecarboxylate, methyl4-bromopicolinate, 2-pyridinecarboxylic acid, 4-bromo-, methyl ester, 4-bromopyridine-2-carboxylic acid methyl ester, abbypharma ap-10-5912, pubchem12733, acmc-209hag, methyl 4-bromo-pyridine-2-carboxylate
JZFLATQBIPILFS-UHFFFAOYSA-N
methyl 4-bromopyridine-2-carboxylate
12828947
≥97%
Methyl 4-bromopyridine-2-carboxylate
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
SDS
Documents
Documents
Product Certifications
RUO – Research Use Only
Product Content Correction
Product Title
Methyl 4-bromopyridine-2-carboxylate, 97+%, Thermo Scientific Chemicals