missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(chloroformyl)butyrate, 98%, Thermo Scientific Chemicals

Product Code. 15299616
Click to view available options
10g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1501-26-4
Density 1.1910g/mL
Boiling Point 110.0°C (17.0 mmHg)
Flash Point 82°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H9ClO3
Linear Formula ClCO(CH2)3CO2CH3
Refractive Index 1.4435 to 1.4455
MDL Number MFCD00000756
Quantity 10 g
Specific Gravity 1.191
Synonym methyl 4-chloroformyl butyrate, methyl 5-chloro-5-oxovalerate, methyl 4-chloroformylbutyrate, methyl5-chloro-5-oxopentanoate, 5-chloro-5-oxopentanoic acid methyl ester, methyl glutaryl chloride, pentanoic acid, 5-chloro-5-oxo-, methyl ester, 4-chlorocarbonyl-butyric acid methyl ester, zlchem 559, pubchem14033
InChI Key JCAZSWWHFJVFPP-UHFFFAOYSA-N
SMILES COC(=O)CCCC(Cl)=O
IUPAC Name methyl 5-chloro-5-oxopentanoate
Molecular Weight (g/mol) 164.59
PubChem CID 73916
Formula Weight 164.59
Percent Purity 98%
Chemical Name or Material Methyl 4-(chloroformyl)butyrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-(chloroformyl)butyrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'