missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4'-methylbiphenyl-4-carboxylate, 95%, Thermo Scientific™

510.00 SEK - 1874.00 SEK

Chemical Identifiers

CAS 49742-56-5
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD05979484
InChI Key NSUWJKRQGXBISO-UHFFFAOYSA-N
Synonym methyl 4'-methylbiphenyl-4-carboxylate, methyl 4'-methyl 1,1'-biphenyl-4-carboxylate, methyl 4-4-methylphenyl benzoate, methyl 4-p-tolyl benzoate, 1,1'-biphenyl-4-carboxylic acid, 4'-methyl-, methyl ester, 4'-methyl-biphenyl-4-carboxylic acid methyl ester, methyl 4'-methyl-1,1'-biphenyl-4-carboxylateShow MoreShow Less
PubChem CID 170805
IUPAC Name methyl 4-(4-methylphenyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)C1=CC=C(C)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15498397
SDS View Documents
Thermo Scientific Alfa Aesar
H51911.03
1 g N/A N/A N/A
15408407
SDS View Documents
Thermo Scientific Alfa Aesar
H51911.06
5 g N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 256-454-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 4'-methylbiphenyl-4-carboxylate, 95%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.