Learn More
Methyl carbamate, 99%, Thermo Scientific Chemicals
334.90 SEK - 1550.22 SEK
Chemical Identifiers
CAS | 598-55-0 |
---|---|
Molecular Formula | C2H5NO2 |
Molecular Weight (g/mol) | 75.07 |
MDL Number | MFCD00007964 |
InChI Key | GTCAXTIRRLKXRU-UHFFFAOYSA-N |
Synonym | urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech |
PubChem CID | 11722 |
ChEBI | CHEBI:76606 |
IUPAC Name | methyl carbamate |
SMILES | COC(N)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10451481
|
Thermo Scientific Acros
269310050 |
5 g | Glass Bottle |
334.90 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10329200
|
Thermo Scientific Acros
269312500 |
250 g | Plastic Bottle |
549.09 SEK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13458498
|
Thermo Scientific Acros
269310010 |
1 kg | Plastic Bottle |
1550.22 SEK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
598-55-0 | |
75.07 | |
GTCAXTIRRLKXRU-UHFFFAOYSA-N | |
11722 | |
methyl carbamate |
C2H5NO2 | |
MFCD00007964 | |
urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech | |
CHEBI:76606 | |
COC(N)=O |
Specifications
54.0°C to 56.0°C | |
176.0°C to 177.0°C | |
99% | |
Authentic | |
MFCD00007964 | |
15, 6110 | |
Solubility in water: 700g/L (20°C). Other solubilities: very soluble in ethanol,soluble in acetone and ether | |
COC(N)=O | |
75.07 | |
CHEBI:76606 | |
99% | |
Adhering Crystals |
598-55-0 | |
White | |
C2H5NO2 | |
H2NCO2CH3 | |
03, 21 | |
urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech | |
GTCAXTIRRLKXRU-UHFFFAOYSA-N | |
methyl carbamate | |
11722 | |
75.06 | |
93°C | |
Methyl carbamate, 99% |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-939-2
missing translation for 'rtecsNumber' : FC2450000
missing translation for 'tsca' : TSCA
RUO – Research Use Only