missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl difluoroacetate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
A methyl difluoroacetate (MFA)-based LiPF6 solution was applied as an electrolyte to improve the thermal stability of Li-ion batteries. Methyl difluoroacetate is used in the inhibition of corrosion of aluminum cathode current collector for lithium ion cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A methyl difluoroacetate (MFA)-based LiPF6 solution was applied as an electrolyte to improve the thermal stability of Li-ion batteries. Methyl difluoroacetate is used in the inhibition of corrosion of aluminum cathode current collector for lithium ion cells.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
A methyl difluoroacetate (MFA)-based LiPF6 solution was applied as an electrolyte to improve the thermal stability of Li-ion batteries. Methyl difluoroacetate is used in the inhibition of corrosion of aluminum cathode current collector for lithium ion cells.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.272 |
Boiling Point | 86°C to 87°C |
Flash Point | 19°C (66°F) |
Refractive Index | 1.335 |
Quantity | 5 g |
UN Number | UN2924 |
Beilstein | 1747534 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 110.06 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'