Learn More
Methyl o-toluate, 99%, Thermo Scientific Chemicals
Product Code. 11467647
Click to view available options
Quantity:
25 g
100 g
500 g
It has been used in the preparation of 2-(azidomethyl)benzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It has been used in the preparation of 2-(azidomethyl)benzoic acid.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89-71-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008428
InChI Key WVWZECQNFWFVFW-UHFFFAOYSA-N
Synonym methyl o-toluate, methyl orthotoluate, o-toluic acid, methyl ester, methyl o-methylbenzoate, methyl toluate, benzoic acid, 2-methyl-, methyl ester, 2-methylbenzoic acid methyl ester, o-toluic acid methyl ester, methyl-o-methyl benzoate, 2-methyl methylbenzoate
PubChem CID 33094
IUPAC Name methyl 2-methylbenzoate
SMILES CC1=CC=CC=C1C(=O)OC
Specifications
Density 1.072
Boiling Point 207°C to 208°C
Flash Point 82°C (179°F)
Refractive Index 1.519
Quantity 25 g
Beilstein 2041846
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Methyl o-toluate