missing translation for 'onlineSavingsMsg'
Learn More

Methyl P-Hydroxybenzoate, MP Biomedicals™

Used as a preservative in foods, beverages and cosmetics. Methyl P-Hydroxybenzoate, MP Biomedicals is the methyl ester of p-hydroxybenzoic acid. It can be used to inhibit yeast growth and can also be used as an antimicrobial agent for Gram-positive anaero

325.00 SEK - 979.00 SEK

Chemical Identifiers

CAS 99-76-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
InChI Key LXCFILQKKLGQFO-UHFFFAOYSA-N
Synonym methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m
PubChem CID 7456
ChEBI CHEBI:31835
IUPAC Name methyl 4-hydroxybenzoate
SMILES COC(=O)C1=CC=C(C=C1)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11403580
View Documents Promotion Details
MP Biomedicals
02102341.1
100 g
325.00 SEK
100g
Please to purchase this item. Need a web account? Register with us today!
11413580
View Documents Promotion Details
MP Biomedicals
02102341.5
500 g
979.00 SEK
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Methyl P-Hydroxybenzoate must be suspended in ethanol first, since it does not dissolve readily in water. It is indicated to have some pheromonal properties..

Specifications

Chemical Identifiers

99-76-3
152.149
methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m
CHEBI:31835
COC(=O)C1=CC=C(C=C1)O
C8H8O3
LXCFILQKKLGQFO-UHFFFAOYSA-N
7456
methyl 4-hydroxybenzoate

Specifications

99-76-3
270°C to 280°C
methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m
LXCFILQKKLGQFO-UHFFFAOYSA-N
methyl 4-hydroxybenzoate
7456
152.1
124°C to 131°C
C8H8O3
Slightly soluble in water (1g/400mL); soluble in warm oil (1g/40mL) or warm glycerol (1g/70mL); freely soluble in alcohol (50mg/mL methanol, clear, colorless solution), acetone or ether.
COC(=O)C1=CC=C(C=C1)O
152.149
CHEBI:31835
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature (15–30°C)

SDS
Documents

Documents

Product Certifications