missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-2-octenoate, 90+%, remainder mainly cis-isomer, Thermo Scientific™

Product Code. 11484157
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7367-81-9
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00036620
InChI Key CJBQSBZJDJHMLF-BQYQJAHWSA-N
Synonym methyl trans-2-octenoate, methyl oct-2-enoate, methyl 2-octenoate, 2-octenoic acid, methyl ester, methyl e-oct-2-enoate, unii-9en9v40j78, 2-octenoic acid, methyl ester, e, 2-octenoic acid, methyl ester, 2e, 2-octenoic acid methyl ester, fema no. 3712
PubChem CID 5364532
IUPAC Name methyl (E)-oct-2-enoate
SMILES CCCCCC=CC(=O)OC

Specifications

Density 0.901
Boiling Point 70°C (4 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.442
Quantity 25 g
Formula Weight 156.22
Percent Purity ≥90%
Assay remainder mainly cis-isomer
Chemical Name or Material Methyl trans-2-octenoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.