Learn More
Methyl 3-(bromomethyl)benzoate, 95%, Thermo Scientific Chemicals
895.31 SEK - 13127.49 SEK
Chemical Identifiers
CAS 1129-28-8
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00051437
InChI Key YUHSMQQNPRLEEJ-UHFFFAOYSA-N
Synonym methyl 3-bromomethyl benzoate, m-carbomethoxybenzyl bromide, 3-bromomethyl benzoic acid methyl ester, benzoic acid, 3-bromomethyl-, methyl ester, methyl-3-bromomethylbenzoate, methyl alpha-bromo-m-toluate, methyl3-bromomethyl benzoate, methyl-3-bromomethyl benzoate, 3-carbomethoxybenzyl bromideShow MoreShow Less
PubChem CID 517981
IUPAC Name methyl 3-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=CC(CBr)=C1
View More Specs
Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10500552
SDS View Documents
Thermo Scientific Acros
368630050
5 g Glass bottle
895.31 SEK
5g
10242622
SDS View Documents
Thermo Scientific Acros
368630250
25 g Glass bottle
3410.19 SEK
25g
10339143
SDS View Documents
Thermo Scientific Acros
368631000
100 g Glass bottle
13127.49 SEK
100g
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Product Certifications
RUO – Research Use Only
Product Content Correction
Product Title
Methyl 3-(bromomethyl)benzoate, 95%, Thermo Scientific Chemicals