Learn More
Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals
4539.30 SEK
Chemical Identifiers
CAS | 512-42-5 |
---|---|
Molecular Formula | CH3NaO4S |
Molecular Weight (g/mol) | 134.08 |
MDL Number | MFCD00013457 |
InChI Key | DZXBHDRHRFLQCJ-UHFFFAOYSA-M |
Synonym | sodium methyl sulfate, methylsulfuric acid sodium salt, methyl sodium sulfate, sodium methyl sulphate, sodium monomethylsulfate, unii-38w76yj3jn, sodium methylsulfate, methyl sulfate sodium salt, methyl sulfate, sodium salt, sulfuric acid, monomethyl ester, sodium salt |
PubChem CID | 2735086 |
IUPAC Name | sodium;methyl sulfate |
SMILES | [Na+].COS([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10677454
|
Thermo Scientific Acros
414901000 |
100 g | Glass bottle |
4539.30 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
512-42-5 | |
134.08 | |
DZXBHDRHRFLQCJ-UHFFFAOYSA-M | |
2735086 | |
[Na+].COS([O-])(=O)=O |
CH3NaO4S | |
MFCD00013457 | |
sodium methyl sulfate, methylsulfuric acid sodium salt, methyl sodium sulfate, sodium methyl sulphate, sodium monomethylsulfate, unii-38w76yj3jn, sodium methylsulfate, methyl sulfate sodium salt, methyl sulfate, sodium salt, sulfuric acid, monomethyl ester, sodium salt | |
sodium;methyl sulfate |
Specifications
512-42-5 | |
100.0 | |
White | |
99% | |
CH3OSO3Na | |
11, 8595 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol | |
[Na+].COS([O-])(=O)=O | |
134.08 | |
134.09 | |
Practical | |
Methylsulfuric acid sodium salt |
98.5 | |
91°C to 94°C | |
Authentic | |
CH3NaO4S | |
MFCD00013457 | |
sodium methyl sulfate, methylsulfuric acid sodium salt, methyl sodium sulfate, sodium methyl sulphate, sodium monomethylsulfate, unii-38w76yj3jn, sodium methylsulfate, methyl sulfate sodium salt, methyl sulfate, sodium salt, sulfuric acid, monomethyl ester, sodium salt | |
DZXBHDRHRFLQCJ-UHFFFAOYSA-M | |
sodium;methyl sulfate | |
2735086 | |
99% | |
Powder |
Safety and Handling
GHS H Statement:
May cause cancer.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-142-7
- Methylsulfuric acid sodium salt
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H350-May cause cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only