Learn More
Metro™ MetroMax i™ Polymer Lab Worktable, Gray Phenolic Top with 3-Sided Frame
Keep your lab cleaner with the Metro MetroMax i™ Polymer Lab Worktable, featuring superior corrosion resistance with built-in Microban™ antimicrobial product protection.
Brand:  Metro™ LTM60XUPG3
Additional Details : Weight : 50.08000kg
 View more versions of this product
Product Code. 15987081
  • 15935.00 SEK / Each
Estimated Shipment: 17-09-2024
to see stock.
Add to basket
Description
Description
These worktables provide the ideal product platform, are modular by design and offer ultimate flexibility based on the application requirements.
  • Advanced polymer construction for corrosive lab environments
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters and accessories must be ordered separately.
Specifications
Specifications
59.75 in.
59.75 x 30 x 40.5 in. (151.7 x 76.2 x 101.6 cm)
101.6 cm
3 Sided frame, adaptor insert
273 kg
151.7 cm
Polymer
Mobile Table
Ideal for lab
40.5 in.
30 in.
76.2 cm
600 lb.