Learn More
Metro™ Stainless Lab Worktable, Black Phenolic Top and 3-Sided Frame
Protect your workers in the laboratory. Metro Stainless Lab Worktable helps to keep them safe with a phenolic top and corrosion-proof, stainless steel construction throughout the body.
Brand:  Metro™ LTSM60UPB
Additional Details : Weight : 43.54400kg
 View more versions of this product
Product Code. 15917151
  • 22810.00 SEK / Each
Estimated Shipment: 17-09-2024
to see stock.
Add to basket
Description
Description
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Specifications
59.75 in.
59.75 x 30 x 35.8 in. (151.7 x 76.2 x 91.1 cm)
91.1 cm
3 Sided frame, adaptor insert
273 kg
151.7 cm
Stainless Steel
Mobile Table
Ideal for lab
35.8 in.
30 in.
76.2 cm
600 lb.