Learn More
Metro™ Stainless Lab Worktable, Black Phenolic Top and 3-Sided Frame
Protect your workers in the laboratory. Metro Stainless Lab Worktable helps to keep them safe with a phenolic top and corrosion-proof, stainless steel construction throughout the body.
Brand:  Metro™ LTS36UPB
Additional Details : Weight : 29.75200kg
 View more versions of this product
Product Code. 15937101
  • 19625.00 SEK / Each
Estimated Shipment: 21-10-2024
to see stock.
Add to basket
Description
Description
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Specifications
35.75 in.
35.75 x 30 x 36 in. (90.8 x 76.2 x 91.4 cm)
91.4 cm
3 Sided frame
273 kg
90.8 cm
Stainless Steel
Stationary Table
Ideal for lab
36 in.
30 in.
76.2 cm
600 lb.