Learn More
Metro™ Stainless Lab Worktable, Black Phenolic Top and Solid HD Shelf
Safeguard your workers in the laboratory. Metro Stainless Lab Worktable with a solid HD shelf and a stainless top/backsplash provides corrosion-proof performance and stainless steel construction throughout.
Brand:  Metro™ LTSM30PB
Additional Details : Weight : 27.94400kg
 View more versions of this product
Product Code. 15957121
  • 18905.00 SEK / Each
Estimated Shipment: 22-10-2024
to see stock.
Add to basket
Description
Description
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Specifications
29.75 in.
29.75 x 30 x 36 in. (74.9 x 76.2 x 91.4 cm)
91.4 cm
Solid HD shelf, adaptor insert
273 kg
74.9 cm
Stainless Steel
Mobile Table
Ideal for lab
36 in.
30 in.
76.2 cm
600 lb.