Learn More
Mettler Toledo™ Self-Adhesive Paper Rolls for Mettler Toledo Balance Printers
For use with Mettler Toledo LC-P43 Printer, BT-P42 (wireless) Printer, and RS-P42 Printers. Mettler Toledo™ Self-Adhesive Paper Rolls for Mettler Toledo Balance Printers are replacement adhesive papers.
Brand: Mettler Toledo™ 11600388
301.99 SEK valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
For use with Mettler Toledo LC-P43 Printer, BT-P42 (wireless) Printer, and RS-P42 Printers. Mettler Toledo™ Self-Adhesive Paper Rolls for Mettler Toledo Balance Printers are replacement adhesive papers.
- Replacement adhesive paper for use with a variety of Mettler Balance printers.
Specifications
Mettler Toledo LC-P43 and the BT-P42 (wireless) | |
3 Pack | |
Paper Roll, Adhesive |
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now