missing translation for 'onlineSavingsMsg'
Learn More

MFF Rabbit anti-Human, Mouse, Rat, Clone: 5U5S4, Novus Biologicals™

Rabbit Monoclonal Antibody

1905.00 SEK - 4805.00 SEK

Specifications

Antigen MFF
Clone 5U5S4
Dilution Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 to 4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18337233
View Documents
Bio-Techne
NBP3-16683-20UL
20 μg
1905.00 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
18389915
View Documents
Bio-Techne
NBP3-16683-100UL
100 μg
4805.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MFF Monoclonal antibody specifically detects MFF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

MFF
Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 to 4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Monoclonal
Purified
RUO
Human, Mouse, Rat
mitochondrial fission factor
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human MFF (Q9GZY8).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
5U5S4
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis, Cell Biology, Endocrinology
PBS, 0.05% BSA, 50% glycerol, pH7.3
56947
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.