missing translation for 'onlineSavingsMsg'
Learn More

(±)-Miconazole, Nitrate, MP Biomedicals™

Product Code. 11442442
Click to view available options
100g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 75319-48-1
Color White
Molecular Formula C18H15Cl4N3O4
Quantity 100 g
Synonym miconazole nitrate, albistat, andergin, aflorix, conofite, florid, micatin, gyno-monistat, epi-monistat, gyno-daktar
InChI Key MCCACAIVAXEFAL-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)Cl)COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl.[N+](=O)(O)[O-]
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
Molecular Weight (g/mol) 479.135
PubChem CID 68553
Formula Weight 479.1
Physical Form Powder
Chemical Name or Material (±)-Miconazole
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.