Learn More
Biochrom Micro-Volume Cuvette
Sample volumes as low as 0.5 μL can be used in the Micro-Volume Cuvette. Samples are pipetted directly onto the sample window meaning more light is transmitted through to the sample which increases measurement range as well as increasing accuracy, unlike other micro-volume cuvettes.
Brand:  Biochrom 80-7100-71
 View more versions of this product
Product Code. 15354108
  • 29635.00 SEK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Exceptional measurement accuracy
  • Wide measurement range - no more changing pathlengths
  • Use only 1 μL of preicous sample
  • Simple to use and clean
  • Easy to check for sample impurities such as bubbles or dust
  • Compatible with most standatd UV-Visible Sprectrophotometer
Specifications
Specifications
Micro-Volume Cuvette with 0.2 mm Pathlength for 15 mm Beam Height Spectrophotometers
12.5 x 12.5 x 61.0 mm
Reusable
15 mm Bean Hight Spectrophotometers
Minimum 0.8 μL
1.2 ng/μL DNA
Microvolume Cuvette
Life Science
0.2 mm
Rectangular