Learn More
Hettich Lab Technology™ Microlitre Centrifuge
The Microlitre Centrifuge can process very small samples at very high speeds. The powerful, maintenance free motors achieve high RCF values for best sample results. Reliable and steady temperature control, quick pre-cool and stand-by cooling functions maintain a constant sample temperature.
Brand:  Hettich Lab Technology™ 2205
 View more versions of this product
Product Code. 10792314
  • 72230.00 SEK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Better results and more samples in less time - increase efficiency and throughput in your lab
  • Wide range of capabilities due to the variety of models and accessories available
  • Pre-cooling function for fast operation - reliable and steady temperature control systems with quick pre-cool and standby functions for precise temperature control
Specifications
Specifications
60 x 1.5/2 mL
31,514 x g
Microlitre Centrifuge
Lid dropping protection
MIKRO 220R
18,000 rpm
-20°C to +40°C
Angle Rotor
650 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products