Learn More
Sartorius Stedim Biotech™ Microsart™ Combi.Jet Traditional 2-Branch Manifold
Sartorius Stedim Biotech™ Microsart™ Combi.Jet Traditional 2-Branch Manifold is constructed of high-grade stainless steel.
Brand: Sartorius Stedim Biotech™ 16847-CJ
This item is not returnable.
View return policy
Description
- Compact design saves valuable laboratory space.
- The Microsart™ Combi.jet is a 2-branch manifold, made of high-grade stainless steel
- Specifically designed for the use together with the new Microsart™ e.jet Transfer Pump (ISO 8199 compliant).
- Creates sufficient vacuum for vacuum filtration concurrently transferring the filtered liquid directly to waste.
- Microsart™ Combi.jet and Microsart™ e.jet can be easily connected and disassembled by the innovative Quick Connection technology
Specifications
Autoclavable (134°C) | |
e.jet transfer pump 166MP-4, 47mm membrane filter | |
2 filter holders with base support and without sterile venting |
For microbial enumeration | |
Stainless Steel |