missing translation for 'onlineSavingsMsg'
Learn More

Motic™ 3-Position Phase Slider for Ph1, Ph2, BF

Combines with phase contrast objectives to observe transparent samples. Motic™ 3-Position Phase Slider for Ph1, Ph2, BF is used with BA Series Microscopes.

Brand:  Motic™ 1101001100531

Product Code. 11732308

  • 1010.00 SEK / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For phase contrast and Bright Field applications.
  • For use with BA Series Microscopes
Specifications

Specifications

Phase Slider
Ph1, Ph2, BF phase -contrast microscopy
BA Series Microscopes
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.