missing translation for 'onlineSavingsMsg'
Learn More

Zeiss™ Microscope Stand for Axio Vert.A1 FL-LED

Features carrier transmitted light illumination for HAL or LED

Brand:  Zeiss™ 431030-9040-000

Product Code. 12362013

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • 5-position nosepiece, H DIC M27 manual (2x H, 3x H DIC) with 10 x 29mm slider mount
  • Carrier transmitted-light illumination for HAL or LED with 2-position filter slider, d: 32mm
  • Reflected-light illuminator FL-LED
  • 4-position mount for LED modules
  • 4-position reflector turret man. for P&C modules
  • Manual coarse/fine focus drive
  • 13mm focus lift with adjustable focus stop
  • RL/TL switching
  • Ergonomic intermediate piece H: 50mm
  • Integrated 12V DC 60W power unit, stabilized, 100 to 240VAC; 50 to 60Hz; 80VA
  • Country-specific power cable
Specifications

Specifications

Microscope Stand for Axio Vert.A1 FL-LED
Axio Vert.A1 FL-LED
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.