missing translation for 'onlineSavingsMsg'
Learn More

ML 323

Potent and selective USP1-UAF1 allosteric inhibitor

788.00 SEK - 2620.00 SEK

Specifications

CAS 1572414-83-5
Synonym 5-Methyl-2-[2-(1-methylethyl)phenyl]-N-[[4-(1H-1,2,3-triazol-1-yl)phenyl]methyl]-4-pyrimidinamine
Molecular Formula C23H24N6
Formula Weight 384.48
Purity 98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18781408
View Documents Promotion Details
Tocris Bioscience
6435/5
5 mg
788.00 SEK
5mg
Please to purchase this item. Need a web account? Register with us today!
18701748
View Documents Promotion Details
Tocris Bioscience
6435/25
25 mg
2620.00 SEK
25mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent and selective allosteric inhibitor of USP1-UAF1 deubiquitinase complex (IC50 = 76 nM). Exhibits selectivity for USP1-UAF1 over a panel of enzymes, including other deuquitinating enzymes. Inhibits deubiquitination of PCNA and FANCD2 and sensitizes cisplatin-resistant H596 cells to cell death induced by cisplatin (Cat. No. 2251) in vitro.
Specifications

Specifications

1572414-83-5
C23H24N6
98%
5-Methyl-2-[2-(1-methylethyl)phenyl]-N-[[4-(1H-1,2,3-triazol-1-yl)phenyl]methyl]-4-pyrimidinamine
384.48
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications