Learn More
MMAF-d8 hydrochloride, MedChemExpress
MedChemExpress D8-MMAF hydrochloride is a deuterated form of MMAF hydrochloride, which is a microtubule disrupting agent.
Brand:  Medchem Express HY-15579AS
Encompass_Preferred
 View more versions of this product
Product Code. 16471529
  • 16400.88 SEK / 1mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
D8-MMAF hydrochloride is a deuterated form of MMAF hydrochloride, which is a microtubule disrupting agent.
Specifications
Specifications
MMAF-d8 hydrochloride
Off-White
C39H58D8ClN5O8
CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C(O)=O)CC2=CC=CC=C2)=O)OC)=O)OC)N(C([C@@](C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C.Cl
776.47
Research
Solid
1 mg
Research
DMSO : 100 mg/mL (128.79 mM; Need ultrasonic)
776.47
98.97%
Cancer-programmed cell death
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.