missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOBS, 0.2M buffer soln., pH 7.5, Thermo Scientific™
Click to view available options
Quantity:
100 mL
250 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. Usage above 20 mM in mammalian cell culture work is not recommended.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Keep away from oxidizing agents.
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. Usage above 20 mM in mammalian cell culture work is not recommended.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Keep away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Concentration | 0.2 M |
pH | 7.5 |
Physical Form | Liquid |
Quantity | 100 mL |
Solubility Information | Soluble in water. |
IUPAC Name | 4-(morpholin-4-yl)butane-1-sulfonic acid |
Chemical Name or Material | MOBS buffer soln. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'