missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum Disulfide, MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
CAS | 1317-33-5 |
Quantity | 100 g |
Molecular Formula | MoS2 |
Synonym | molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z |
InChI Key | CWQXQMHSOZUFJS-UHFFFAOYSA-N |
SMILES | S=[Mo]=S |
IUPAC Name | disulfanylidenemolybdenum |
Molecular Weight (g/mol) | 160.07 |
PubChem CID | 14823 |
ChEBI | CHEBI:30704 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'