Learn More
Motic™ Moticam A16
The ultimate Moticam for advanced enthusiasts with demanding needs, the Moticam A16 has a large 1/2 in. sCMOS sensor with resolution above 4K and delivers up to 15 fps in live imaging.
Brand: Motic™ 1100600101463
Description
- C-mount for lens
- MotiConnect software is available for free download (with free updates included)
- Live Display Modes: 4608 x 3456 px, 3264 x 2448 px, 1920 x 1080 px
- Maximum Frame Rate: 4608 x 3456 at 15 fps, 3264 x 2448 at 15 fps, 1920 x 1080 at 30 fps
Specifications
Camera | |
Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap | |
1/2.3 in. |
Moticam A | |
sCMOS | |
15 to 30 fps |