missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Angiopoietin-1 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB8220
This item is not returnable.
View return policy
Description
Angiopoietin-1 Monoclonal specifically detects Angiopoietin-1 in Mouse samples. It is validated for Western Blot.Specifications
Angiopoietin-1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, ANGPT1, KIAA0003 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse Angiopoietin-1 in ELISA and Western Blot. In ELISA, no cross-reactivity with recombinant mouse Angiopoietin-2 is detected. | |
Mouse | |
Purified |
Western Blot | |
892732 | |
Unconjugated | |
O08538 | |
ANGPT1 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Angiopoietin-1 Ser20-Phe498 Accession # O08538 | |
100 μg | |
284 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |