Learn More
Fc gamma RI/CD64 Mouse anti-Human, Clone: 10.1, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
Brand:  R&D Systems MAB1257-500
Encompass
Product Code. 15793954
This item is not returnable. View return policy
Description
Description
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with rheumatoid synovial fluid cells and fibronectin purified monocytes. The IgG fraction of the tissue culture supernatant was purified by Protein G affinity chromatography.
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Fc gamma RI/CD64 Mouse anti-Human, Clone: 10.1, R&D Systems™